Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291508
Encompass
HR2-915-08/Crystal Screen 2 Cryo #08 (E08), 185 ml - 1.05 M Sodium chloride, 7% v/v Ethanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-5