Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Catalog No. 501587
Encompass
HR2-915-30/Crystal Screen 2 Cryo #30 (G06), 185 ml - 0.08 M HEPES pH 7.5, 8% w/v Polyethylene glycol 6,000, 4% v/v (+/-)-2-Methyl-2,4-pentanediol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-7 Supplier Hampton Research Supplier No. HR291530
HR2-915-30/Crystal Screen 2 Cryo #30 (G06), 185 ml - 0.08 M HEPES pH 7.5, 8% w/v Polyethylene glycol 6,000, 4% v/v (+/-)-2-Methyl-2,4-pentanediol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.