Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291505
Encompass
HR2-915-05/Crystal Screen 2 Cryo #05 (E05), 185 ml - 1.5 M Ammonium sulfate, 3.75% v/v 2-Propanol, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-2