Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291513
Encompass
HR2-915-13/Crystal Screen 2 Cryo #13 (F01), 185 ml - 0.18 M Ammonium sulfate, 0.09 M Sodium acetate trihydrate pH 4.6, 27% w/v Polyethylene glycol monomethyl ether 2,000, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-0