Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291542
Encompass
HR2-915-42/Crystal Screen 2 Cryo #42 (H06), 185 ml - 1.275 M Ammonium sulfate, 0.085 M Tris pH 8.5, 25.2% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-9