Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291527
Encompass
HR2-915-27/Crystal Screen 2 Cryo #27 (G03), 185 ml - 0.009 M Zinc sulfate heptahydrate, 0.09 M MES monohydrate pH 6.5, 22.5% v/v Polyethylene glycol monomethyl ether 550, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-4