Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291539
Encompass
HR2-915-39/Crystal Screen 2 Cryo #39 (H03), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Tris pH 8.5, 3.4 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-6