Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291518
Encompass
HR2-915-18/Crystal Screen 2 Cryo #18 (F06), 185 ml - 0.007 M Iron(III) chloride hexahydrate, 0.07 M Sodium citrate tribasic dihydrate pH 5.6, 7% v/v Jeffamine M-600, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-5