Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291541
Encompass
HR2-915-41/Crystal Screen 2 Cryo #41 (H05), 185 ml - 0.0075 M Nickel(II) chloride hexahydrate, 0.075 M Tris pH 8.5, 0.75 M Lithium sulfate monohydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-8