Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291533
Encompass
HR2-915-33/Crystal Screen 2 Cryo #33 (G09), 185 ml - 0.07 M HEPES pH 7.5, 1.4 M Ammonium formate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-0