Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291514
Encompass
HR2-915-14/Crystal Screen 2 Cryo #14 (F02), 185 ml - 0.15 M Potassium sodium tartrate tetrahydrate, 0.075 M Sodium citrate tribasic dihydrate pH 5.6, 1.5 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-1