Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291504
Encompass
HR2-915-04/Crystal Screen 2 Cryo #04 (E04), 185 ml - 26.25% v/v 1,4-Dioxane, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-1