Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291536
Encompass
HR2-915-36/Crystal Screen 2 Cryo #36 (G12), 185 ml - 0.085 M HEPES pH 7.5, 3.655 M Sodium chloride, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-3