Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291525
Encompass
HR2-915-25/Crystal Screen 2 Cryo #25 (G01), 185 ml - 0.0075 M Cobalt(II) chloride hexahydrate, 0.075 M MES monohydrate pH 6.5, 1.35 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-2