Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291512
Encompass
HR2-915-12/Crystal Screen 2 Cryo #12 (E12), 185 ml - 0.095 M Cadmium chloride hydrate, 0.095 M Sodium acetate trihydrate pH 4.6, 28.5% v/v Polyethylene glycol 400, 5% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-9