Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291546
Encompass
HR2-915-46/Crystal Screen 2 Cryo #46 (H10), 185 ml - 0.085 M Sodium chloride, 0.085 M BICINE pH 9.0, 17% v/v Polyethylene glycol monomethyl ether 550, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-3