Learn More
Hampton Research CRYSTAL SCREEN 2 NO01 185ML
SDP
Catalog No. 501460
Encompass
HR2-912-01/Crystal Screen 2 #01 (E01), 185 ml - 2.0 M Sodium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-146-0 Supplier Hampton Research Supplier No. HR291201
HR2-912-01/Crystal Screen 2 #01 (E01), 185 ml - 2.0 M Sodium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.