Learn More
Hampton Research CRYSTAL SCREEN 2 NO05 185ML
SDP
Catalog No. 501464
Encompass
HR2-912-05/Crystal Screen 2 #05 (E05), 185 ml - 2.0 M Ammonium sulfate, 5% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-146-4 Supplier Hampton Research Supplier No. HR291205
HR2-912-05/Crystal Screen 2 #05 (E05), 185 ml - 2.0 M Ammonium sulfate, 5% v/v 2-Propanol. Please note items from this supplier are non-returnable.