Learn More
Hampton Research CRYSTAL SCREEN 2 NO06 185ML
SDP
Catalog No. 501465
Encompass
HR2-912-06/Crystal Screen 2 #06 (E06), 185 ml - 1.0 M Imidazole pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-146-5 Supplier Hampton Research Supplier No. HR291206
HR2-912-06/Crystal Screen 2 #06 (E06), 185 ml - 1.0 M Imidazole pH 7.0. Please note items from this supplier are non-returnable.