Learn More
Hampton Research CRYSTAL SCREEN 2 NO07 185ML
SDP
Catalog No. 501466
Encompass
HR2-912-07/Crystal Screen 2 #07 (E07), 185 ml - 10% w/v Polyethylene glycol 1,000, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-146-6 Supplier Hampton Research Supplier No. HR291207
HR2-912-07/Crystal Screen 2 #07 (E07), 185 ml - 10% w/v Polyethylene glycol 1,000, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.