Learn More
Hampton Research CRYSTAL SCREEN 2 NO10 185ML
SDP
Supplier:  Hampton Research HR291210
Encompass
HR2-912-10/Crystal Screen 2 #10 (E10), 185 ml - 0.2 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-146-9