Learn More
Hampton Research CRYSTAL SCREEN 2 NO11 185ML
SDP
Catalog No. 501470
Encompass
HR2-912-11/Crystal Screen 2 #11 (E11), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-147-0 Supplier Hampton Research Supplier No. HR291211
HR2-912-11/Crystal Screen 2 #11 (E11), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.