Learn More
Hampton Research CRYSTAL SCREEN 2 NO12 185ML
SDP
Catalog No. 501471
Encompass
HR2-912-12/Crystal Screen 2 #12 (E12), 185 ml - 0.1 M Cadmium chloride hydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-147-1 Supplier Hampton Research Supplier No. HR291212
HR2-912-12/Crystal Screen 2 #12 (E12), 185 ml - 0.1 M Cadmium chloride hydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.