Learn More
Hampton Research CRYSTAL SCREEN 2 NO13 185ML
SDP
Catalog No. 501472
Encompass
HR2-912-13/Crystal Screen 2 #13 (F01), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-147-2 Supplier Hampton Research Supplier No. HR291213
HR2-912-13/Crystal Screen 2 #13 (F01), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.