Learn More
Hampton Research CRYSTAL SCREEN 2 NO14 185ML
SDP
Catalog No. 501473
Encompass
HR2-912-14/Crystal Screen 2 #14 (F02), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-147-3 Supplier Hampton Research Supplier No. HR291214
HR2-912-14/Crystal Screen 2 #14 (F02), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.