Learn More
Hampton Research CRYSTAL SCREEN 2 NO15 185ML
SDP
Catalog No. 501474
Encompass
HR2-912-15/Crystal Screen 2 #15 (F03), 185 ml - 0.5 M Ammonium sulfate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-147-4 Supplier Hampton Research Supplier No. HR291215
HR2-912-15/Crystal Screen 2 #15 (F03), 185 ml - 0.5 M Ammonium sulfate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.