Learn More
Hampton Research CRYSTAL SCREEN 2 NO17 185ML
SDP
Catalog No. 501476
Encompass
HR2-912-17/Crystal Screen 2 #17 (F05), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 35% v/v tert-Butanol. Please note items from this supplier are non-returnable.
Catalog No. 50-147-6 Supplier Hampton Research Supplier No. HR291217
HR2-912-17/Crystal Screen 2 #17 (F05), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 35% v/v tert-Butanol. Please note items from this supplier are non-returnable.