Learn More
Hampton Research CRYSTAL SCREEN 2 NO18 185ML
SDP
Catalog No. 501477
Encompass
HR2-912-18/Crystal Screen 2 #18 (F06), 185 ml - 0.01 M Iron(III) chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 10% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.
Catalog No. 50-147-7 Supplier Hampton Research Supplier No. HR291218
HR2-912-18/Crystal Screen 2 #18 (F06), 185 ml - 0.01 M Iron(III) chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 10% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.