Learn More
Hampton Research CRYSTAL SCREEN 2 NO19 185ML
SDP
Catalog No. 501478
Encompass
HR2-912-19/Crystal Screen 2 #19 (F07), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 2.5 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-147-8 Supplier Hampton Research Supplier No. HR291219
HR2-912-19/Crystal Screen 2 #19 (F07), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 2.5 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.