Learn More
Hampton Research CRYSTAL SCREEN 2 NO20 185ML
SDP
Catalog No. 501479
Encompass
HR2-912-20/Crystal Screen 2 #20 (F08), 185 ml - 0.1 M MES monohydrate pH 6.5, 1.6 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-147-9 Supplier Hampton Research Supplier No. HR291220
HR2-912-20/Crystal Screen 2 #20 (F08), 185 ml - 0.1 M MES monohydrate pH 6.5, 1.6 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.