Learn More
Hampton Research CRYSTAL SCREEN 2 NO21 185ML
SDP
Catalog No. 501480
Encompass
HR2-912-21/Crystal Screen 2 #21 (F09), 185 ml - 0.1 M Sodium phosphate monobasic monohydrate, 0.1 M Potassium phosphate monobasic, 0.1 M MES monohydrate pH 6.5, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-148-0 Supplier Hampton Research Supplier No. HR291221
HR2-912-21/Crystal Screen 2 #21 (F09), 185 ml - 0.1 M Sodium phosphate monobasic monohydrate, 0.1 M Potassium phosphate monobasic, 0.1 M MES monohydrate pH 6.5, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.