Learn More
Hampton Research CRYSTAL SCREEN 2 NO22 185ML
SDP
Catalog No. 501481
Encompass
HR2-912-22/Crystal Screen 2 #22 (F10), 185 ml - 0.1 M MES monohydrate pH 6.5, 12% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-148-1 Supplier Hampton Research Supplier No. HR291222
HR2-912-22/Crystal Screen 2 #22 (F10), 185 ml - 0.1 M MES monohydrate pH 6.5, 12% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.