Learn More
Hampton Research CRYSTAL SCREEN 2 NO23 185ML
SDP
Catalog No. 501482
Encompass
HR2-912-23/Crystal Screen 2 #23 (F11), 185 ml - 1.6 M Ammonium sulfate, 0.1 M MES monohydrate pH 6.5, 10% v/v 1,4-Dioxane. Please note items from this supplier are non-returnable.
Catalog No. 50-148-2 Supplier Hampton Research Supplier No. HR291223
HR2-912-23/Crystal Screen 2 #23 (F11), 185 ml - 1.6 M Ammonium sulfate, 0.1 M MES monohydrate pH 6.5, 10% v/v 1,4-Dioxane. Please note items from this supplier are non-returnable.