Learn More
Hampton Research CRYSTAL SCREEN 2 NO24 185ML
SDP
Catalog No. 501483
Encompass
HR2-912-24/Crystal Screen 2 #24 (F12), 185 ml - 0.05 M Cesium chloride, 0.1 M MES monohydrate pH 6.5, 30% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.
Catalog No. 50-148-3 Supplier Hampton Research Supplier No. HR291224
HR2-912-24/Crystal Screen 2 #24 (F12), 185 ml - 0.05 M Cesium chloride, 0.1 M MES monohydrate pH 6.5, 30% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.