Learn More
Hampton Research CRYSTAL SCREEN 2 NO25 185ML
SDP
Catalog No. 501484
Encompass
HR2-912-25/Crystal Screen 2 #25 (G01), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M MES monohydrate pH 6.5, 1.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-148-4 Supplier Hampton Research Supplier No. HR291225
HR2-912-25/Crystal Screen 2 #25 (G01), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M MES monohydrate pH 6.5, 1.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.