Learn More
Hampton Research CRYSTAL SCREEN 2 NO26 185ML
SDP
Catalog No. 501485
Encompass
HR2-912-26/Crystal Screen 2 #26 (G02), 185 ml - 0.2 M Ammonium sulfate, 0.1 M MES monohydrate pH 6.5, 30% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-148-5 Supplier Hampton Research Supplier No. HR291226
HR2-912-26/Crystal Screen 2 #26 (G02), 185 ml - 0.2 M Ammonium sulfate, 0.1 M MES monohydrate pH 6.5, 30% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.