Learn More
Hampton Research CRYSTAL SCREEN 2 NO27 185ML
SDP
Catalog No. 501486
Encompass
HR2-912-27/Crystal Screen 2 #27 (G03), 185 ml - 0.01 M Zinc sulfate heptahydrate, 0.1 M MES monohydrate pH 6.5, 25% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-148-6 Supplier Hampton Research Supplier No. HR291227
HR2-912-27/Crystal Screen 2 #27 (G03), 185 ml - 0.01 M Zinc sulfate heptahydrate, 0.1 M MES monohydrate pH 6.5, 25% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.