Learn More
Hampton Research CRYSTAL SCREEN 2 NO30 185ML
SDP
Catalog No. 501489
Encompass
HR2-912-30/Crystal Screen 2 #30 (G06), 185 ml - 0.1 M HEPES pH 7.5, 10% w/v Polyethylene glycol 6,000, 5% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-148-9 Supplier Hampton Research Supplier No. HR291230
HR2-912-30/Crystal Screen 2 #30 (G06), 185 ml - 0.1 M HEPES pH 7.5, 10% w/v Polyethylene glycol 6,000, 5% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.