Learn More
Hampton Research CRYSTAL SCREEN 2 NO31 185ML
SDP
Catalog No. 501490
Encompass
HR2-912-31/Crystal Screen 2 #31 (G07), 185 ml - 0.1 M HEPES pH 7.5, 20% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.
Catalog No. 50-149-0 Supplier Hampton Research Supplier No. HR291231
HR2-912-31/Crystal Screen 2 #31 (G07), 185 ml - 0.1 M HEPES pH 7.5, 20% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.