Learn More
Hampton Research CRYSTAL SCREEN 2 NO32 185ML
SDP
Catalog No. 501491
Encompass
HR2-912-32/Crystal Screen 2 #32 (G08), 185 ml - 0.1 M Sodium chloride, 0.1 M HEPES pH 7.5, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-149-1 Supplier Hampton Research Supplier No. HR291232
HR2-912-32/Crystal Screen 2 #32 (G08), 185 ml - 0.1 M Sodium chloride, 0.1 M HEPES pH 7.5, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.