Learn More
Hampton Research CRYSTAL SCREEN 2 NO33 185ML
SDP
Catalog No. 501492
Encompass
HR2-912-33/Crystal Screen 2 #33 (G09), 185 ml - 0.1 M HEPES pH 7.5, 2.0 M Ammonium formate. Please note items from this supplier are non-returnable.
Catalog No. 50-149-2 Supplier Hampton Research Supplier No. HR291233
HR2-912-33/Crystal Screen 2 #33 (G09), 185 ml - 0.1 M HEPES pH 7.5, 2.0 M Ammonium formate. Please note items from this supplier are non-returnable.