Learn More
Hampton Research CRYSTAL SCREEN 2 NO34 185ML
SDP
Catalog No. 501493
Encompass
HR2-912-34/Crystal Screen 2 #34 (G10), 185 ml - 0.05 M Cadmium sulfate hydrate, 0.1 M HEPES pH 7.5, 1.0 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-149-3 Supplier Hampton Research Supplier No. HR291234
HR2-912-34/Crystal Screen 2 #34 (G10), 185 ml - 0.05 M Cadmium sulfate hydrate, 0.1 M HEPES pH 7.5, 1.0 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.