Learn More
Hampton Research CRYSTAL SCREEN 2 NO35 185ML
SDP
Catalog No. 501494
Encompass
HR2-912-35/Crystal Screen 2 #35 (G11), 185 ml - 0.1 M HEPES pH 7.5, 70% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-149-4 Supplier Hampton Research Supplier No. HR291235
HR2-912-35/Crystal Screen 2 #35 (G11), 185 ml - 0.1 M HEPES pH 7.5, 70% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.