Learn More
Hampton Research CRYSTAL SCREEN 2 NO36 185ML
SDP
Catalog No. 501495
Encompass
HR2-912-36/Crystal Screen 2 #36 (G12), 185 ml - 0.1 M HEPES pH 7.5, 4.3 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-149-5 Supplier Hampton Research Supplier No. HR291236
HR2-912-36/Crystal Screen 2 #36 (G12), 185 ml - 0.1 M HEPES pH 7.5, 4.3 M Sodium chloride. Please note items from this supplier are non-returnable.