Learn More
Hampton Research CRYSTAL SCREEN 2 NO37 185ML
SDP
Catalog No. 501496
Encompass
HR2-912-37/Crystal Screen 2 #37 (H01), 185 ml - 0.1 M HEPES pH 7.5, 8% v/v Ethylene glycol, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-149-6 Supplier Hampton Research Supplier No. HR291237
HR2-912-37/Crystal Screen 2 #37 (H01), 185 ml - 0.1 M HEPES pH 7.5, 8% v/v Ethylene glycol, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.