Learn More
Hampton Research CRYSTAL SCREEN 2 NO38 185ML
SDP
Catalog No. 501497
Encompass
HR2-912-38/Crystal Screen 2 #38 (H02), 185 ml - 0.1 M HEPES pH 7.5, 20% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.
Catalog No. 50-149-7 Supplier Hampton Research Supplier No. HR291238
HR2-912-38/Crystal Screen 2 #38 (H02), 185 ml - 0.1 M HEPES pH 7.5, 20% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.