Learn More
Hampton Research CRYSTAL SCREEN 2 NO39 185ML
SDP
Catalog No. 501498
Encompass
HR2-912-39/Crystal Screen 2 #39 (H03), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Tris pH 8.5, 3.4 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-149-8 Supplier Hampton Research Supplier No. HR291239
HR2-912-39/Crystal Screen 2 #39 (H03), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Tris pH 8.5, 3.4 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.