Learn More
Hampton Research CRYSTAL SCREEN 2 NO41 185ML
SDP
Catalog No. 501500
Encompass
HR2-912-41/Crystal Screen 2 #41 (H05), 185 ml - 0.01 M Nickel(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-150-0 Supplier Hampton Research Supplier No. HR291241
HR2-912-41/Crystal Screen 2 #41 (H05), 185 ml - 0.01 M Nickel(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.