Learn More
Hampton Research CRYSTAL SCREEN 2 NO42 185ML
SDP
Catalog No. 501501
Encompass
HR2-912-42/Crystal Screen 2 #42 (H06), 185 ml - 1.5 M Ammonium sulfate, 0.1 M Tris pH 8.5, 12% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-150-1 Supplier Hampton Research Supplier No. HR291242
HR2-912-42/Crystal Screen 2 #42 (H06), 185 ml - 1.5 M Ammonium sulfate, 0.1 M Tris pH 8.5, 12% v/v Glycerol. Please note items from this supplier are non-returnable.