Learn More
Hampton Research CRYSTAL SCREEN 2 NO43 185ML
SDP
Catalog No. 501502
Encompass
HR2-912-43/Crystal Screen 2 #43 (H07), 185 ml - 0.2 M Ammonium phosphate monobasic, 0.1 M Tris pH 8.5, 50% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-150-2 Supplier Hampton Research Supplier No. HR291243
HR2-912-43/Crystal Screen 2 #43 (H07), 185 ml - 0.2 M Ammonium phosphate monobasic, 0.1 M Tris pH 8.5, 50% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.